Learn More
Meso Scale Discovery UPLEX HUM VEGFR1/Flt1 ASYQP
SDP
Catalog No. 5000080476
Encompass
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a multiplex immunoassay kit that enables simultaneous measurement of VEGF Receptor 1 (VEGFR-1) and other biomarkers in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate quantification of VEGFR-1 levels with high dynamic range and low sample volume requirements The assay has high specificity with no cross-reactivity and includes reagents for 50 tests in one sector of an MSD MULTI-ARRAY 96-well plate
Catalog No. 50-000-80476 Supplier Meso Scale Discovery Supplier No. K151AHHK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a multiplex immunoassay kit that enables simultaneous measurement of VEGF Receptor 1 (VEGFR-1) and other biomarkers in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate quantification of VEGFR-1 levels with high dynamic range and low sample volume requirements The assay has high specificity with no cross-reactivity and includes reagents for 50 tests in one sector of an MSD MULTI-ARRAY 96-well plate