Learn More
Meso Scale Discovery UPLEX HUM VEGFR1/Flt1 ASYQP
SDP
Catalog No. 5000080599
Encompass
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a multiplex immunoassay kit that enables the simultaneous measurement of human Vascular Endothelial Growth Factor Receptor 1 (VEGFR-1) and other biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology providing high sensitivity and a large dynamic range The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for the quantitative determination of VEGFR-1 levels with a lower limit of detection of 19 pg/mL
Catalog No. 50-000-80599 Supplier Meso Scale Discovery Supplier No. K151AHHK21
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a multiplex immunoassay kit that enables the simultaneous measurement of human Vascular Endothelial Growth Factor Receptor 1 (VEGFR-1) and other biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology providing high sensitivity and a large dynamic range The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for the quantitative determination of VEGFR-1 levels with a lower limit of detection of 19 pg/mL