Learn More
Meso Scale Discovery UPLEX HUM VEGFR1/FltASY384
SDP
Catalog No. 5000080274
Encompass
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of human Vascular Endothelial Growth Factor Receptor 1 (VEGFR-1) in cell culture supernatants serum plasma and other biological matrices The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results This product allows for the accurate quantification of VEGFR-1 levels with as little as 25 L of sample
Catalog No. 50-000-80274 Supplier Meso Scale Discovery Supplier No. K251AHHK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of human Vascular Endothelial Growth Factor Receptor 1 (VEGFR-1) in cell culture supernatants serum plasma and other biological matrices The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and rapid results This product allows for the accurate quantification of VEGFR-1 levels with as little as 25 L of sample