Learn More
Meso Scale Discovery UPLEX HUM vWF ASY384well
SDP
Catalog No. 5000078380
Encompass
The Meso Scale Discovery U-PLEX Human VWF 384-Well Assay Kit is a multiplex immunoassay for measuring von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity and broad dynamic range The kit includes a VWF-specific antibody coated plate detection antibody calibrator and optimized assay diluent With a streamlined workflow and small sample volume it provides a sensitive and reproducible tool for studying VWF biology and disease mechanisms
Catalog No. 50-000-78380 Supplier Meso Scale Discovery Supplier No. K251C9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human VWF 384-Well Assay Kit is a multiplex immunoassay for measuring von Willebrand Factor (VWF) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity and broad dynamic range The kit includes a VWF-specific antibody coated plate detection antibody calibrator and optimized assay diluent With a streamlined workflow and small sample volume it provides a sensitive and reproducible tool for studying VWF biology and disease mechanisms