Learn More
Meso Scale Discovery UPLEX HUM YKL40 ASY384well
SDP
Catalog No. 5000080617
Encompass
The YKL-40 ELISA Kit from Meso Scale Discovery is an in vitro diagnostic tool for quantifying YKL-40 in serum plasma and other fluids It uses electrochemiluminescence for high sensitivity and rapid results The assay is a quantitative sandwich ELISA with monoclonal antibodies specific to YKL-40 measuring 3 16-10 000 pg/mL with no cross-reactivity with other chitinase family members The kit includes a 96-well plate antibodies standards substrate and instructions aiding in monitoring diseases like asthma COPD liver fibrosis and certain cancers
Catalog No. 50-000-80617 Supplier Meso Scale Discovery Supplier No. K251VLK4
Add to Cart
Add to Cart
Request a Quote
The YKL-40 ELISA Kit from Meso Scale Discovery is an in vitro diagnostic tool for quantifying YKL-40 in serum plasma and other fluids It uses electrochemiluminescence for high sensitivity and rapid results The assay is a quantitative sandwich ELISA with monoclonal antibodies specific to YKL-40 measuring 3 16-10 000 pg/mL with no cross-reactivity with other chitinase family members The kit includes a 96-well plate antibodies standards substrate and instructions aiding in monitoring diseases like asthma COPD liver fibrosis and certain cancers