Learn More
Meso Scale Discovery U-PLEX Human alpha-Synuclein Kit, 1 plate
SDP
Catalog No. NC1210677
Encompass
The U-PLEX Human alpha-Synuclein Kit has the essential components for an assay that measures alpha-Synuclein in a single-analyte format. It is validated for the measurement of alpha-Synuclein in human cerebrospinal fluid, plasma, saliva, and serum.
Catalog No. NC1210677 Supplier Meso Scale Discovery Supplier No. K151WKK1
The U-PLEX Human alpha-Synuclein Kit has the essential components for an assay that measures alpha-Synuclein in a single-analyte format. It is validated for the measurement of alpha-Synuclein in human cerebrospinal fluid, plasma, saliva, and serum.