Learn More
Meso Scale Discovery U-PLEX Human Eotaxin-3 Assay, 384-well format, Single-analyte, Mouse monoclonal antibodies, LLOD 7.3 pg/mL, Dynamic range 7.3-21,400 pg/mL
SDP
Catalog No. NC2741767
Encompass
U-PLEX Human Eotaxin-3 Assay, 384-well format, Single-analyte, Mouse monoclonal antibodies, LLOD 7.3 pg/mL, Dynamic range 7.3-21,400 pg/mL
Catalog No. NC2741767 Supplier Meso Scale Discovery Supplier No. K251UEK2
Add to Cart
Add to Cart
Request a Quote
U-PLEX Human Eotaxin-3 Assay, 384-well format, Single-analyte, Mouse monoclonal antibodies, LLOD 7.3 pg/mL, Dynamic range 7.3-21,400 pg/mL