Learn More
Meso Scale Discovery U-PLEX Human FGF-21 Assay, QuickPlex (1 PL)
SDP
Catalog No. NC3786843
Encompass
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human FGF-21 Assay has the essential components for an assay that measures FGF-21 in a single-analyte format. For information on multiplexing FGF-21 with other analytes, please see U-PLEX U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:Fibroblast Growth Factor 21, mitogen, morphogenesis, metabolic diseases, gluconeogenesis, ketogenesis, diabetes"
Catalog No. NC3786843 Supplier Meso Scale Discovery Supplier No. K1515WK21
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human FGF-21 Assay has the essential components for an assay that measures FGF-21 in a single-analyte format. For information on multiplexing FGF-21 with other analytes, please see U-PLEX U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:Fibroblast Growth Factor 21, mitogen, morphogenesis, metabolic diseases, gluconeogenesis, ketogenesis, diabetes"