Learn More
Meso Scale Discovery U-PLEX Human FGF-23 Assay, SECTOR (5 PL)
SDP
Catalog No. NC3786733
Encompass
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Assay has the essential components for an assay that measures single-analyte format. For information on multiplexing with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:Glucagon-Like Peptide-1, GLP-1, metabolic diseases, metabolism, proglucagon, insulin, diabetes"
Catalog No. NC3786733 Supplier Meso Scale Discovery Supplier No. K1516EK2
Add to Cart
Add to Cart
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Assay has the essential components for an assay that measures single-analyte format. For information on multiplexing with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:Glucagon-Like Peptide-1, GLP-1, metabolic diseases, metabolism, proglucagon, insulin, diabetes"