Learn More
Meso Scale Discovery U-PLEX Human Ghrelin (total) Assay, QuickPlex (25 PL)
SDP
Catalog No. NC3786853
Encompass
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human Ghrelin (total) Assay has the essential components for an assay that measures Ghrelin (total) in a single-analyte format. For information on multiplexing Ghrelin (total) with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:GHRL, MTLRP, obestatin, GHSR, obesity"
Catalog No. NC3786853 Supplier Meso Scale Discovery Supplier No. K1515XK24
Add to Cart
Add to Cart
Request a Quote
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human Ghrelin (total) Assay has the essential components for an assay that measures Ghrelin (total) in a single-analyte format. For information on multiplexing Ghrelin (total) with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:GHRL, MTLRP, obestatin, GHSR, obesity"