Learn More
Meso Scale Discovery U-PLEX Human GIP (total) Assay, SECTOR (25 PL)
SDP
Catalog No. NC3786645
Encompass
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Assay has the essential components for an assay that measures single-analyte format. For information on multiplexing with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:Glucagon-Like Peptide-1, GLP-1, metabolic diseases, metabolism, proglucagon, insulin, diabetes"
Catalog No. NC3786645 Supplier Meso Scale Discovery Supplier No. K1515SK4
Add to Cart
Add to Cart
Request a Quote
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Assay has the essential components for an assay that measures single-analyte format. For information on multiplexing with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:Glucagon-Like Peptide-1, GLP-1, metabolic diseases, metabolism, proglucagon, insulin, diabetes"