Learn More
Meso Scale Discovery U-PLEX Human GIP (total) Assay, QuickPlex (25 PL)
SDP
Catalog No. NC3786829
Encompass
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human GIP (total) Assay has the essential components for an assay that measures GIP (total) in a single-analyte format. For information on multiplexing GIP (total) with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:Gastric Inhibitory Polypeptide, Glucose-dependent Insulinotropic Polypeptide, GIP (1-42), incretin, metabolic diseases, metabolism, homeostasis, obesity, diabetes"
Catalog No. NC3786829 Supplier Meso Scale Discovery Supplier No. K1515SK24
Add to Cart
Add to Cart
Request a Quote
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human GIP (total) Assay has the essential components for an assay that measures GIP (total) in a single-analyte format. For information on multiplexing GIP (total) with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:Gastric Inhibitory Polypeptide, Glucose-dependent Insulinotropic Polypeptide, GIP (1-42), incretin, metabolic diseases, metabolism, homeostasis, obesity, diabetes"