Learn More
Meso Scale Discovery U-PLEX Human Glucagon Assay, QuickPlex (25 PL)
SDP
Catalog No. NC3786861
Encompass
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human Glucagon Assay has the essential components for an assay that measures Glucagon in a single-analyte format. For information on multiplexing Glucagon with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:GCG, metabolism, metabolic, gluconeogenesis, glycogenolysis, ketogenesis, glucose, Metabolic Syndrome, diabetes"
Catalog No. NC3786861 Supplier Meso Scale Discovery Supplier No. K1515YK24
Add to Cart
Add to Cart
Request a Quote
"The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. U-PLEX Human Glucagon Assay has the essential components for an assay that measures Glucagon in a single-analyte format. For information on multiplexing Glucagon with other analytes, please see U-PLEX Custom Metabolic Group 1 (human) Assays. Related Key Terms:GCG, metabolism, metabolic, gluconeogenesis, glycogenolysis, ketogenesis, glucose, Metabolic Syndrome, diabetes"