Learn More
Meso Scale Discovery U-PLEX Human IL-23 Assay, 25 plate size, IL-23 analyte, Human species reactivity, 96-well plate type, 1.4-21,600 pg/mL dynamic range, Serum, Plasma, Cell Culture Supernatant sample types
SDP
Catalog No. NC2944432
Encompass
U-PLEX Human IL-23 Assay, 25 plate size, IL-23 analyte, Human species reactivity, 96-well plate type, 1.4-21,600 pg/mL dynamic range, Serum, Plasma, Cell Culture Supernatant sample types
Catalog No. NC2944432 Supplier Meso Scale Discovery Supplier No. K151WGK4
Add to Cart
Add to Cart
Request a Quote
U-PLEX Human IL-23 Assay, 25 plate size, IL-23 analyte, Human species reactivity, 96-well plate type, 1.4-21,600 pg/mL dynamic range, Serum, Plasma, Cell Culture Supernatant sample types