Learn More
Meso Scale Discovery U-PLEX Human TARC Assay, 384-well, Single-analyte, Sensitivity 0.51 pg/mL, Dynamic Range 0.51-2200 pg/mL, Kit Size 25 plates, Research Use Only
SDP
Catalog No. NC2394162
Encompass
U-PLEX Human TARC Assay, 384-well, Single-analyte, Sensitivity 0.51 pg/mL, Dynamic Range 0.51-2200 pg/mL, Kit Size 25 plates, Research Use Only
Catalog No. NC2394162 Supplier Meso Scale Discovery Supplier No. K251ULK4
Add to Cart
Add to Cart
Request a Quote
U-PLEX Human TARC Assay, 384-well, Single-analyte, Sensitivity 0.51 pg/mL, Dynamic Range 0.51-2200 pg/mL, Kit Size 25 plates, Research Use Only