Learn More
Meso Scale Discovery UPLEX Inflammation Combo 1 hu
SDP
Catalog No. 5000080346
Encompass
The Meso Scale Discovery U-PLEX Inflammation Combo 1 Human Kit is a multiplex immunoassay for measuring IL-1 IL-6 IL-8 IL-10 IL-12p70 IL-13 IFN- and TNF- in human serum plasma and cell culture supernatants It uses MSD s MULTI-ARRAY technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies read buffer calibrators and controls for quantitative analysis of 8 biomarkers ideal for biomarker profiling immunology and drug discovery research
Catalog No. 50-000-80346 Supplier Meso Scale Discovery Supplier No. K15387K1
The Meso Scale Discovery U-PLEX Inflammation Combo 1 Human Kit is a multiplex immunoassay for measuring IL-1 IL-6 IL-8 IL-10 IL-12p70 IL-13 IFN- and TNF- in human serum plasma and cell culture supernatants It uses MSD s MULTI-ARRAY technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies read buffer calibrators and controls for quantitative analysis of 8 biomarkers ideal for biomarker profiling immunology and drug discovery research