Learn More
Meso Scale Discovery UPLEX Inflammation Combo hu
SDP
Catalog No. 5000081369
Encompass
The Meso Scale Discovery U-PLEX Inflammation Combo 1 Human Kit is a multiplex immunoassay that allows simultaneous quantitative measurement of IL-1 IL-6 IL-8 IL-10 IL-12p70 IL-13 IFN- and TNF- biomarkers It uses electrochemiluminescence detection technology and is designed for use with human serum plasma and cell culture supernatant samples The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators It allows sensitive reproducible results from small sample volumes of 25-50 L
Catalog No. 50-000-81369 Supplier Meso Scale Discovery Supplier No. K15387K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Inflammation Combo 1 Human Kit is a multiplex immunoassay that allows simultaneous quantitative measurement of IL-1 IL-6 IL-8 IL-10 IL-12p70 IL-13 IFN- and TNF- biomarkers It uses electrochemiluminescence detection technology and is designed for use with human serum plasma and cell culture supernatant samples The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators It allows sensitive reproducible results from small sample volumes of 25-50 L