Learn More
Meso Scale Discovery UPLEX Kidney Injury Combo hu
SDP
Catalog No. 5000079809
Encompass
The Meso Scale Discovery U-PLEX Kidney Injury Combo 1 Human Kit is a multiplex immunoassay that simultaneously measures human biomarkers for kidney injury and damage This research kit includes reagents to run the assay on the MSD platform and quantitatively measures KIM-1 NGAL Cystatin C Clusterin and MCP-1 in human serum plasma urine and cell culture supernatants The electrochemiluminescent detection allows for high sensitivity and a wide dynamic range This kit is for research use only and not intended for diagnostic procedures It provides a streamlined workflow for assessing kidney toxicity and injury
Catalog No. 50-000-79809 Supplier Meso Scale Discovery Supplier No. K15389K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Kidney Injury Combo 1 Human Kit is a multiplex immunoassay that simultaneously measures human biomarkers for kidney injury and damage This research kit includes reagents to run the assay on the MSD platform and quantitatively measures KIM-1 NGAL Cystatin C Clusterin and MCP-1 in human serum plasma urine and cell culture supernatants The electrochemiluminescent detection allows for high sensitivity and a wide dynamic range This kit is for research use only and not intended for diagnostic procedures It provides a streamlined workflow for assessing kidney toxicity and injury