Learn More
Meso Scale Discovery UPLEX Kidney Injury Combo hu
SDP
Catalog No. 5000081167
Encompass
The Meso Scale Discovery U-PLEX Kidney Injury Combo 1 Human Kit (K15389K) is a multiplex immunoassay that measures biomarkers of kidney injury and function It includes reagents to measure human KIM-1 NGAL Cystatin C and Creatinine in serum plasma urine and cell culture supernatants The electrochemiluminescent detection allows sensitive and accurate quantification of the biomarkers This kit can aid in early detection of acute kidney injury and assessment of kidney function The kit contains sufficient reagents for one 96-well plate when used as directed
Catalog No. 50-000-81167 Supplier Meso Scale Discovery Supplier No. K15389K1
The Meso Scale Discovery U-PLEX Kidney Injury Combo 1 Human Kit (K15389K) is a multiplex immunoassay that measures biomarkers of kidney injury and function It includes reagents to measure human KIM-1 NGAL Cystatin C and Creatinine in serum plasma urine and cell culture supernatants The electrochemiluminescent detection allows sensitive and accurate quantification of the biomarkers This kit can aid in early detection of acute kidney injury and assessment of kidney function The kit contains sufficient reagents for one 96-well plate when used as directed