Learn More
Meso Scale Discovery UPLEX Kidney Injury Combo hu
SDP
Catalog No. 5000082292
Encompass
The Meso Scale Discovery U-PLEX Kidney Injury Combo 1 Human Kit (K15389K) is a multiplex immunoassay that measures biomarkers of kidney injury and function It includes reagents for 96 tests to quantitatively measure human KIM-1 NGAL Cystatin C and Creatinine in serum plasma urine and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements It can aid in the assessment of acute kidney injury and chronic kidney disease
Catalog No. 50-000-82292 Supplier Meso Scale Discovery Supplier No. K15389K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Kidney Injury Combo 1 Human Kit (K15389K) is a multiplex immunoassay that measures biomarkers of kidney injury and function It includes reagents for 96 tests to quantitatively measure human KIM-1 NGAL Cystatin C and Creatinine in serum plasma urine and cell culture supernatants This assay kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements It can aid in the assessment of acute kidney injury and chronic kidney disease