Learn More
Meso Scale Discovery UPLEX Macrophage M2 Combo hu
SDP
Catalog No. 5000079845
Encompass
The U-PLEX Macrophage M2 Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of human IL-10 CCL17 CCL18 and CCL22 in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer and calibrator The electrochemiluminescent assay takes 3 5 hours to complete and has a detection range of 0 05-2500 pg/mL This kit allows high-throughput sensitive quantification of M2 macrophage activation markers
Catalog No. 50-000-79845 Supplier Meso Scale Discovery Supplier No. K15337K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Macrophage M2 Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of human IL-10 CCL17 CCL18 and CCL22 in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer and calibrator The electrochemiluminescent assay takes 3 5 hours to complete and has a detection range of 0 05-2500 pg/mL This kit allows high-throughput sensitive quantification of M2 macrophage activation markers