Learn More
Meso Scale Discovery UPLEX Macrophage M2 Combo ms
SDP
Catalog No. 5000078232
Encompass
The Meso Scale Discovery U-PLEX Combinations are multiplex immunoassay panels that allow simultaneous detection of up to 10 protein biomarkers from a single sample The assays utilize electrochemiluminescent detection for high sensitivity and broad dynamic range The U-PLEX panels are available in several combinations targeting cytokines chemokines growth factors and other soluble proteins They require as little as 25-50 L of sample and can be run on the MSD QuickPlex SQ120 MESO QuickPlex SQ120 or SECTOR Imager 2400 platforms The assays provide quantitative results in as little as 3 5 hours
Catalog No. 50-000-78232 Supplier Meso Scale Discovery Supplier No. K15409K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Combinations are multiplex immunoassay panels that allow simultaneous detection of up to 10 protein biomarkers from a single sample The assays utilize electrochemiluminescent detection for high sensitivity and broad dynamic range The U-PLEX panels are available in several combinations targeting cytokines chemokines growth factors and other soluble proteins They require as little as 25-50 L of sample and can be run on the MSD QuickPlex SQ120 MESO QuickPlex SQ120 or SECTOR Imager 2400 platforms The assays provide quantitative results in as little as 3 5 hours