Learn More
Meso Scale Discovery UPLEX Macrophage MCombo ms
SDP
Catalog No. 5000080205
Encompass
The U-PLEX Macrophage M1 Combo 1 by Meso Scale Diagnostics is a multiplex assay kit for measuring human IFN-gamma IL-12p70 IL-1beta IL-6 TNF-alpha and IL-8 It uses electrochemiluminescent detection on Meso Scale platforms The assay forms a sandwich complex with monoclonal antibodies immobilized on carbon electrodes Light emitted is proportional to analyte levels The kit includes a 96-well plate calibrators controls diluent detection antibodies buffers seals and instructions enabling sensitive quantification of inflammatory biomarkers from cell culture supernatants
Catalog No. 50-000-80205 Supplier Meso Scale Discovery Supplier No. K15408K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Macrophage M1 Combo 1 by Meso Scale Diagnostics is a multiplex assay kit for measuring human IFN-gamma IL-12p70 IL-1beta IL-6 TNF-alpha and IL-8 It uses electrochemiluminescent detection on Meso Scale platforms The assay forms a sandwich complex with monoclonal antibodies immobilized on carbon electrodes Light emitted is proportional to analyte levels The kit includes a 96-well plate calibrators controls diluent detection antibodies buffers seals and instructions enabling sensitive quantification of inflammatory biomarkers from cell culture supernatants