Learn More
Meso Scale Discovery U-PLEX Metabolic 2-Plex Combo 1 (hu) SECTOR (1 PL)
SDP
Catalog No. NC3786819
Encompass
U-PLEX Combos offer popular panels of biomarkers that have been grouped together for convenient ordering. They have been tested for multiplex compatibility and are fulfilled as a kit. The U-PLEX Metabolic Combo is an extensive combination of assays for human metabolic biomarkers. The assays can be used for quantitative measurements in the sample types listed in the "Specifications" tab below, and offer detection limits (LLODs) in the sub- to low pg/mL range.
Catalog No. NC3786819 Supplier Meso Scale Discovery Supplier No. K15282K1
Add to Cart
Add to Cart
U-PLEX Combos offer popular panels of biomarkers that have been grouped together for convenient ordering. They have been tested for multiplex compatibility and are fulfilled as a kit. The U-PLEX Metabolic Combo is an extensive combination of assays for human metabolic biomarkers. The assays can be used for quantitative measurements in the sample types listed in the "Specifications" tab below, and offer detection limits (LLODs) in the sub- to low pg/mL range.