Learn More
Meso Scale Discovery UPLEX Metabolic 2Plex 2 rat
SDP
Catalog No. 5000079614
Encompass
The U-PLEX Metabolic 2-Plex Combo 2 Rat kit from Meso Scale Diagnostics measures insulin and leptin in rat serum plasma and cell culture supernatants using electrochemiluminescent detection It offers high sensitivity a broad dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and diluent It supports diabetes obesity and energy regulation research with a 3 5-hour protocol and minimal hands-on time
Catalog No. 50-000-79614 Supplier Meso Scale Discovery Supplier No. K15313K1
The U-PLEX Metabolic 2-Plex Combo 2 Rat kit from Meso Scale Diagnostics measures insulin and leptin in rat serum plasma and cell culture supernatants using electrochemiluminescent detection It offers high sensitivity a broad dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and diluent It supports diabetes obesity and energy regulation research with a 3 5-hour protocol and minimal hands-on time