Learn More
Meso Scale Discovery UPLEX Metabolic 2Plex 2 rat
SDP
Catalog No. 5000079638
Encompass
The U-PLEX Metabolic 2-Plex Combo 2 Rat is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin and leptin in rat serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results with low sample volume requirements This 2-plex panel enables efficient analysis of metabolic biomarkers related to energy regulation and obesity research in rat models The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents sufficient for one 96-well plate
Catalog No. 50-000-79638 Supplier Meso Scale Discovery Supplier No. K15313K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic 2-Plex Combo 2 Rat is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin and leptin in rat serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results with low sample volume requirements This 2-plex panel enables efficient analysis of metabolic biomarkers related to energy regulation and obesity research in rat models The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents sufficient for one 96-well plate