Learn More
Meso Scale Discovery UPLEX Metabolic 2Plex 2
SDP
Catalog No. 5000079612
Encompass
The U-PLEX Metabolic 2-plex Combo 2 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin and leptin in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The assay requires 50 L sample volume and can be read on MESO QuickPlex SQ 120 QuickPlex SQ 120 or QuickPlex SQ 240 instruments The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls It allows sensitive reproducible and convenient analysis of insulin and leptin from mouse samples
Catalog No. 50-000-79612 Supplier Meso Scale Discovery Supplier No. K15303K2
The U-PLEX Metabolic 2-plex Combo 2 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin and leptin in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The assay requires 50 L sample volume and can be read on MESO QuickPlex SQ 120 QuickPlex SQ 120 or QuickPlex SQ 240 instruments The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls It allows sensitive reproducible and convenient analysis of insulin and leptin from mouse samples