Learn More
Meso Scale Discovery UPLEX Metabolic 2Plex 2
SDP
Catalog No. 5000081872
Encompass
The U-PLEX Metabolic 2-plex Combo 2 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin and leptin in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The assay requires 50 L of sample and can be read on Meso Scale Discovery instruments capable of SECTOR plate imaging This 2-plex kit provides reagents for one 96-well plate which can test 40 samples in duplicate
Catalog No. 50-000-81872 Supplier Meso Scale Discovery Supplier No. K15303K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic 2-plex Combo 2 Mouse is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin and leptin in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The assay requires 50 L of sample and can be read on Meso Scale Discovery instruments capable of SECTOR plate imaging This 2-plex kit provides reagents for one 96-well plate which can test 40 samples in duplicate