Learn More
Meso Scale Discovery UPLEX Metabolic 2Plex
SDP
Catalog No. 5000079639
Encompass
The U-PLEX Metabolic 2-plex Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of insulin and leptin in mouse samples The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay The electrochemiluminescent detection allows for high sensitivity and a broad dynamic range This kit is designed for research use only to study metabolic diseases in mouse models The kit has enough reagents to run one 96-well plate testing 40 samples in duplicate
Catalog No. 50-000-79639 Supplier Meso Scale Discovery Supplier No. K15302K2
The U-PLEX Metabolic 2-plex Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of insulin and leptin in mouse samples The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay The electrochemiluminescent detection allows for high sensitivity and a broad dynamic range This kit is designed for research use only to study metabolic diseases in mouse models The kit has enough reagents to run one 96-well plate testing 40 samples in duplicate