Learn More
Meso Scale Discovery UPLEX Metabolic 2Plex rat
SDP
Catalog No. 5000080413
Encompass
The U-PLEX Metabolic 2-plex Combo 1 Rat kit by Meso Scale Diagnostics measures insulin and leptin in rat serum plasma and tissue culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for simultaneous analyte measurement in one well It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents The kit offers a dynamic range of 0 05-1000 pg/mL for insulin and 0 2-2000 pg/mL for leptin with results in under 5 hours
Catalog No. 50-000-80413 Supplier Meso Scale Discovery Supplier No. K15312K2
The U-PLEX Metabolic 2-plex Combo 1 Rat kit by Meso Scale Diagnostics measures insulin and leptin in rat serum plasma and tissue culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for simultaneous analyte measurement in one well It includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents The kit offers a dynamic range of 0 05-1000 pg/mL for insulin and 0 2-2000 pg/mL for leptin with results in under 5 hours