Learn More
Meso Scale Discovery UPLEX Metabolic 3Plex 2 rat
SDP
Catalog No. 5000082130
Encompass
The U-PLEX Metabolic 3-plex Combo 2 Rat is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin leptin and resistin in rat serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements This 3-plex panel enables researchers to efficiently profile key metabolic biomarkers related to energy regulation and homeostasis in rat models With a short 2 5 hour protocol it is well-suited for studies involving large sample sets
Catalog No. 50-000-82130 Supplier Meso Scale Discovery Supplier No. K15315K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic 3-plex Combo 2 Rat is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of insulin leptin and resistin in rat serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements This 3-plex panel enables researchers to efficiently profile key metabolic biomarkers related to energy regulation and homeostasis in rat models With a short 2 5 hour protocol it is well-suited for studies involving large sample sets