Learn More
Meso Scale Discovery UPLEX Metabolic 3Plex 2 rat
SDP
Catalog No. 5000082483
Encompass
The U-PLEX Metabolic 3-Plex Combo 2 Rat is a multiplex immunoassay kit from Meso Scale Diagnostics for measuring insulin leptin and resistin in rat serum plasma and tissue culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume This 3-plex panel efficiently analyzes metabolic biomarkers related to energy regulation and glucose homeostasis in rat models The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and reagents for one 96-well plate
Catalog No. 50-000-82483 Supplier Meso Scale Discovery Supplier No. K15315K1
The U-PLEX Metabolic 3-Plex Combo 2 Rat is a multiplex immunoassay kit from Meso Scale Diagnostics for measuring insulin leptin and resistin in rat serum plasma and tissue culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume This 3-plex panel efficiently analyzes metabolic biomarkers related to energy regulation and glucose homeostasis in rat models The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and reagents for one 96-well plate