Learn More
Meso Scale Discovery UPLEX Metabolic 3Plex 2
SDP
Catalog No. 5000079613
Encompass
The U-PLEX Metabolic 3-Plex Combo 2 is a multiplex immunoassay kit from Meso Scale Diagnostics for measuring insulin leptin and resistin in serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and diluent It efficiently analyzes metabolic biomarkers related to obesity diabetes and insulin resistance with a 4 5-hour protocol and minimal hands-on time
Catalog No. 50-000-79613 Supplier Meso Scale Discovery Supplier No. K15305K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic 3-Plex Combo 2 is a multiplex immunoassay kit from Meso Scale Diagnostics for measuring insulin leptin and resistin in serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and diluent It efficiently analyzes metabolic biomarkers related to obesity diabetes and insulin resistance with a 4 5-hour protocol and minimal hands-on time