Learn More
Meso Scale Discovery UPLEX Metabolic 3Plex 2
SDP
Catalog No. 5000082391
Encompass
The Meso Scale Discovery U-PLEX Combinations are multiplex immunoassays that allow simultaneous detection of up to 10 protein biomarkers in human samples The assays utilize electrochemiluminescent detection for high sensitivity and broad dynamic range The U-PLEX platform provides a streamlined workflow from sample to answer with minimal hands-on time There are over 40 U-PLEX assay panels available covering inflammation neurobiology oncology and cardiovascular research areas The assays are available in several kit sizes and sample types including serum plasma CSF and cell culture supernatants
Catalog No. 50-000-82391 Supplier Meso Scale Discovery Supplier No. K15305K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Combinations are multiplex immunoassays that allow simultaneous detection of up to 10 protein biomarkers in human samples The assays utilize electrochemiluminescent detection for high sensitivity and broad dynamic range The U-PLEX platform provides a streamlined workflow from sample to answer with minimal hands-on time There are over 40 U-PLEX assay panels available covering inflammation neurobiology oncology and cardiovascular research areas The assays are available in several kit sizes and sample types including serum plasma CSF and cell culture supernatants