Learn More
Meso Scale Discovery UPLEX Metabolic 3Plex 2
SDP
Catalog No. 5000082485
Encompass
The Meso Scale Discovery U-PLEX Combinations are multiplex immunoassay panels that allow simultaneous detection of up to 10 protein biomarkers from a single sample The assays utilize electrochemiluminescent detection for high sensitivity and broad dynamic range The U-PLEX panels are available for human mouse rat monkey canine and porcine samples They enable efficient and reproducible analysis of biomarkers for applications such as immunology oncology neurobiology and cardiovascular research The assays are run on Meso Scale Discovery instruments including QuickPlex MESO QuickPlex SQ 120 and SECTOR Imager 2400
Catalog No. 50-000-82485 Supplier Meso Scale Discovery Supplier No. K15305K1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Combinations are multiplex immunoassay panels that allow simultaneous detection of up to 10 protein biomarkers from a single sample The assays utilize electrochemiluminescent detection for high sensitivity and broad dynamic range The U-PLEX panels are available for human mouse rat monkey canine and porcine samples They enable efficient and reproducible analysis of biomarkers for applications such as immunology oncology neurobiology and cardiovascular research The assays are run on Meso Scale Discovery instruments including QuickPlex MESO QuickPlex SQ 120 and SECTOR Imager 2400