Learn More
Meso Scale Discovery UPLEX Metabolic Combo 1 ms
SDP
Catalog No. 5000078453
Encompass
The U-PLEX Metabolic Combo 1 Mouse Assay from Meso Scale Diagnostics is a multiplex immunoassay panel for quantifying insulin leptin resistin and PAI-1 in mouse samples like serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SECTOR technology offering high sensitivity broad dynamic range low sample volume and a 2 5-hour turnaround The kit includes a 96-well plate with capture antibodies calibrators detection antibodies read buffer wash buffer and plate seals for one 96-well plate enabling efficient metabolic biomarker profiling in mice
Catalog No. 50-000-78453 Supplier Meso Scale Discovery Supplier No. K15297K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic Combo 1 Mouse Assay from Meso Scale Diagnostics is a multiplex immunoassay panel for quantifying insulin leptin resistin and PAI-1 in mouse samples like serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SECTOR technology offering high sensitivity broad dynamic range low sample volume and a 2 5-hour turnaround The kit includes a 96-well plate with capture antibodies calibrators detection antibodies read buffer wash buffer and plate seals for one 96-well plate enabling efficient metabolic biomarker profiling in mice