Learn More
Meso Scale Discovery UPLEX Metabolic Combo 2 hu
SDP
Catalog No. 5000079808
Encompass
The U-PLEX Metabolic Combo 2 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of insulin leptin PAI-1 active resistin and TNF-alpha in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay takes approximately 4 hours to complete and has a detection range of 0 05-2500 pg/mL across the analytes This quantitative immunoassay provides high sensitivity and requires low sample volume
Catalog No. 50-000-79808 Supplier Meso Scale Discovery Supplier No. K15388K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic Combo 2 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of insulin leptin PAI-1 active resistin and TNF-alpha in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay takes approximately 4 hours to complete and has a detection range of 0 05-2500 pg/mL across the analytes This quantitative immunoassay provides high sensitivity and requires low sample volume