Learn More
Meso Scale Discovery UPLEX Metabolic Hormone
SDP
Catalog No. 5000082131
Encompass
The U-PLEX Metabolic Hormones Combo 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of insulin leptin and resistin in rat serum plasma and tissue culture media The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit allows efficient analysis of metabolic and energy homeostasis biomarkers related to obesity diabetes and metabolic syndrome in rat samples
Catalog No. 50-000-82131 Supplier Meso Scale Discovery Supplier No. K15316K1
The U-PLEX Metabolic Hormones Combo 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of insulin leptin and resistin in rat serum plasma and tissue culture media The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit allows efficient analysis of metabolic and energy homeostasis biomarkers related to obesity diabetes and metabolic syndrome in rat samples