Learn More
Meso Scale Discovery UPLEX Metabolic Hormone
SDP
Catalog No. 5000082484
Encompass
The U-PLEX Metabolic Hormones Combo 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of insulin leptin and resistin in rat serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The electrochemiluminescent assay allows simultaneous measurement of the 3 analytes from a single sample in under 3 hours using an MSD instrument The kit has a broad dynamic range and provides accurate and reproducible results for metabolic and diabetes research in rat models
Catalog No. 50-000-82484 Supplier Meso Scale Discovery Supplier No. K15316K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic Hormones Combo 1 Rat Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of insulin leptin and resistin in rat serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The electrochemiluminescent assay allows simultaneous measurement of the 3 analytes from a single sample in under 3 hours using an MSD instrument The kit has a broad dynamic range and provides accurate and reproducible results for metabolic and diabetes research in rat models