Learn More
Meso Scale Discovery UPLEX Metabolic Hormone
SDP
Catalog No. 5000078681
Encompass
The Meso Scale Discovery U-PLEX Metabolic Hormones Combo 1 Mouse Assay Kit is a multiplex immunoassay measuring insulin leptin and resistin in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible quantification from small samples The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents It aids in understanding energy regulation useful in obesity diabetes and metabolic syndrome research
Catalog No. 50-000-78681 Supplier Meso Scale Discovery Supplier No. K15306K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Metabolic Hormones Combo 1 Mouse Assay Kit is a multiplex immunoassay measuring insulin leptin and resistin in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible quantification from small samples The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents It aids in understanding energy regulation useful in obesity diabetes and metabolic syndrome research