Learn More
Meso Scale Discovery UPLEX Metabolic Hormone
SDP
Catalog No. 5000082163
Encompass
The U-PLEX Metabolic Hormones Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring insulin leptin and resistin in mouse samples It includes a 96-well plate with capture antibodies detection antibodies calibrators and buffers The electrochemiluminescent detection method offers high sensitivity and a broad dynamic range Suitable for cell culture supernatants serum plasma and other samples it requires 25 L per well providing a simple and fast workflow to quantify hormone levels
Catalog No. 50-000-82163 Supplier Meso Scale Discovery Supplier No. K15306K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metabolic Hormones Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring insulin leptin and resistin in mouse samples It includes a 96-well plate with capture antibodies detection antibodies calibrators and buffers The electrochemiluminescent detection method offers high sensitivity and a broad dynamic range Suitable for cell culture supernatants serum plasma and other samples it requires 25 L per well providing a simple and fast workflow to quantify hormone levels