Learn More
Meso Scale Discovery UPLEX Metastasis Combo 1HMN
SDP
Catalog No. 5000078225
Encompass
The U-PLEX Metastasis Combo 1 Human Sector Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 protein biomarkers related to metastasis in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies for the 10 analytes detection antibodies read buffer wash buffer diluent calibrators and controls The 10-plex panel includes VEGF-C VEGF-D HGF uPA PAI-1 MMP-2 MMP-3 MMP-9 TIMP-1 and TIMP-2 The electrochemiluminescent assay takes 3 5 hours to complete and has a detection range from 0 01-10 000 pg/mL depending on the analyte
Catalog No. 50-000-78225 Supplier Meso Scale Discovery Supplier No. K15602K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Metastasis Combo 1 Human Sector Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 protein biomarkers related to metastasis in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies for the 10 analytes detection antibodies read buffer wash buffer diluent calibrators and controls The 10-plex panel includes VEGF-C VEGF-D HGF uPA PAI-1 MMP-2 MMP-3 MMP-9 TIMP-1 and TIMP-2 The electrochemiluminescent assay takes 3 5 hours to complete and has a detection range from 0 01-10 000 pg/mL depending on the analyte