Learn More
Meso Scale Discovery UPLEX Metastasis Combo 1HMN
SDP
Catalog No. 5000081446
Encompass
The U-PLEX Metastasis Combo 1 Human Sector Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 protein biomarkers related to metastasis in human serum plasma and cell culture supernatants The kit contains a 10-plex plate detection antibodies calibrators and ancillary reagents needed to run the assay The 10 analytes measured are CCL2 CCL7 CXCL1 CXCL5 CXCL8 CXCL12 IL6 IL8 MMP2 and MMP9 This kit allows high-throughput sensitive and reproducible quantification of multiple metastasis biomarkers from a small sample volume
Catalog No. 50-000-81446 Supplier Meso Scale Discovery Supplier No. K15602K1
The U-PLEX Metastasis Combo 1 Human Sector Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 protein biomarkers related to metastasis in human serum plasma and cell culture supernatants The kit contains a 10-plex plate detection antibodies calibrators and ancillary reagents needed to run the assay The 10 analytes measured are CCL2 CCL7 CXCL1 CXCL5 CXCL8 CXCL12 IL6 IL8 MMP2 and MMP9 This kit allows high-throughput sensitive and reproducible quantification of multiple metastasis biomarkers from a small sample volume