Learn More
Meso Scale Discovery UPLEX Mouse 6CKine/CCL21 ASYQP
SDP
Catalog No. 5000081327
Encompass
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of 6 mouse cytokines and chemokines (CCL21 GM-CSF IFN- IL-1 IL-6 and TNF- ) from cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes all necessary reagents and diluents to perform the assay The sensitivity ranges from 0 3-4 5 pg/mL depending on the analyte The assay requires a small sample volume and provides high-sensitivity results in just 3 5 hours
Catalog No. 50-000-81327 Supplier Meso Scale Discovery Supplier No. K152C0K22
The Meso Scale Discovery U-PLEX Mouse 6CKine (CCL21) Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of 6 mouse cytokines and chemokines (CCL21 GM-CSF IFN- IL-1 IL-6 and TNF- ) from cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes all necessary reagents and diluents to perform the assay The sensitivity ranges from 0 3-4 5 pg/mL depending on the analyte The assay requires a small sample volume and provides high-sensitivity results in just 3 5 hours