Learn More
Meso Scale Discovery UPLEX Mouse CD40/TNFRSF5 ASYQP
SDP
Catalog No. 5000082105
Encompass
The U-PLEX Mouse CD40 Assay from Meso Scale Discovery is a multiplex immunoassay kit that enables simultaneous quantification of mouse CD40 in cell culture supernatants serum plasma and other biological matrices It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The lower limit of detection is 1 3 pg/mL The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-82105 Supplier Meso Scale Discovery Supplier No. K152D0K21
Add to Cart
Add to Cart
The U-PLEX Mouse CD40 Assay from Meso Scale Discovery is a multiplex immunoassay kit that enables simultaneous quantification of mouse CD40 in cell culture supernatants serum plasma and other biological matrices It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The lower limit of detection is 1 3 pg/mL The assay takes approximately 3 5 hours to complete