Learn More
Meso Scale Discovery UPLEX Mouse FGF21 ASY384well
SDP
Catalog No. 5000078969
Encompass
The Meso Scale Discovery U-PLEX Mouse FGF-21 384-Well Assay Kit is a multiplex immunoassay that measures mouse fibroblast growth factor 21 (FGF-21) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls It allows high throughput screening and only requires 10 L sample volume per well The assay has a typical sensitivity of 19 5 pg/mL and an upper limit of quantification of 50 000 pg/mL
Catalog No. 50-000-78969 Supplier Meso Scale Discovery Supplier No. K2525WK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse FGF-21 384-Well Assay Kit is a multiplex immunoassay that measures mouse fibroblast growth factor 21 (FGF-21) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls It allows high throughput screening and only requires 10 L sample volume per well The assay has a typical sensitivity of 19 5 pg/mL and an upper limit of quantification of 50 000 pg/mL