Learn More
Meso Scale Discovery UPLEX Mouse GLP1 active ASY
SDP
Catalog No. 5000082177
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) Assay Kit K1526LK is an electrochemiluminescent immunoassay for the quantitative determination of active glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with a GLP-1 specific capture antibody and a ruthenylated detection antibody to provide high sensitivity and a broad dynamic range The kit includes a plate detection antibody calibrator and assay diluent valid for 96 tests with a 3 5 hour total assay time
Catalog No. 50-000-82177 Supplier Meso Scale Discovery Supplier No. K1526LK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) Assay Kit K1526LK is an electrochemiluminescent immunoassay for the quantitative determination of active glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with a GLP-1 specific capture antibody and a ruthenylated detection antibody to provide high sensitivity and a broad dynamic range The kit includes a plate detection antibody calibrator and assay diluent valid for 96 tests with a 3 5 hour total assay time