Learn More
Meso Scale Discovery UPLEX Mouse GLP1 total ASYQP
SDP
Catalog No. 5000079384
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total Assay Kit measures total GLP-1 in mouse serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from 25 L samples The kit includes a 96-well plate with capture antibody detection antibody with ruthenium calibrators and necessary reagents Results are read on MSD s QuickPlex SQ 120 and analyzed with MSD Discovery Workbench software offering a rapid high-throughput method for measuring GLP-1
Catalog No. 50-000-79384 Supplier Meso Scale Discovery Supplier No. K1525UK21
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total Assay Kit measures total GLP-1 in mouse serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from 25 L samples The kit includes a 96-well plate with capture antibody detection antibody with ruthenium calibrators and necessary reagents Results are read on MSD s QuickPlex SQ 120 and analyzed with MSD Discovery Workbench software offering a rapid high-throughput method for measuring GLP-1