Learn More
Meso Scale Discovery UPLEX Mouse GLP1 total ASYQP
SDP
Catalog No. 5000081323
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total Assay Kit measures total GLP-1 in mouse serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results It includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD s SECTOR Imager and analyzed with MSD Discovery Workbench software offering a rapid high-throughput method for measuring GLP-1 levels
Catalog No. 50-000-81323 Supplier Meso Scale Discovery Supplier No. K1525UK22
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total Assay Kit measures total GLP-1 in mouse serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results It includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD s SECTOR Imager and analyzed with MSD Discovery Workbench software offering a rapid high-throughput method for measuring GLP-1 levels