Learn More
Meso Scale Discovery UPLEX Mouse GMCSF ASY 25 PL
SDP
Catalog No. 5000079337
Encompass
The Meso Scale Discovery U-PLEX Mouse GM-CSF Assay is a multiplex immunoassay kit for measuring mouse GM-CSF using electrochemiluminescence detection It includes a plate with capture antibodies ECL-tagged detection antibodies and calibrator solutions for a standard curve The assay s sensitivity is 0 19 pg/mL with an upper quantification limit of 10 000 pg/mL Compatible with serum plasma and cell culture supernatant the kit provides reagents for 1 x 96-well plate
Catalog No. 50-000-79337 Supplier Meso Scale Discovery Supplier No. K152UMK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse GM-CSF Assay is a multiplex immunoassay kit for measuring mouse GM-CSF using electrochemiluminescence detection It includes a plate with capture antibodies ECL-tagged detection antibodies and calibrator solutions for a standard curve The assay s sensitivity is 0 19 pg/mL with an upper quantification limit of 10 000 pg/mL Compatible with serum plasma and cell culture supernatant the kit provides reagents for 1 x 96-well plate